Learn More
Aviva Systems Biology Corp Reg1 Antibody - middle region (ARP63516_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63516_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Reg1. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-287-994