Learn More
Aviva Systems Biology Corp REG1A Antibody - C-terminal region (ARP63515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REG1A. It was validated on Western Blot. . Alternate names: ICRF, MGC12447, P19, PSP, PSPS, PSPS1, PTP, REGApplicationWB. Species reactivity: Human
Catalog No. 50-287-993