Learn More
Aviva Systems Biology Corp REG1B Antibody - N-terminal region (ARP34029_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REG1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human REG1B. Alternate names: PSPS2, REGH, REGI-BETA, REGLApplicationWB. Species reactivity: Human
Catalog No. 50-014-55223