Learn More
Aviva Systems Biology Corp REG3A Antibody - N-terminal region (ARP51178_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51178_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REG3A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human REG3A. Alternate names: HIP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-562