Learn More
Sino Biological REG3A Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11235R019100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human REG3A (rh REG3A; Catalog#11235-H08H; Q06141-1; Met 1-Asp 175).
Catalog No. 50-224-7689