Learn More
Sino Biological REG3A / HIP Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11235RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human REG3A (rh REG3A; Catalog#11235-H08H; Q06141-1; Met 1-Asp 175). REG3A specific IgG was purified by human REG3A affinity chromatography.
Catalog No. 50-224-8019