Learn More
ACROBiosystems Recombinant Protein;Human Reg4 Protein;HEK293;100UG;RE4,REG4,GISP,REG-IV,RELP,REG-4
Catalog No. 501658191
Encompass_Preferred
Recombinant Protein;Human Reg4 Protein;100UG;Human Reg4, His Tag (RE4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Pro 158 (Accession # AAH17089).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8191 Supplier ACROBiosystems Supplier No. RE4H5221100UG
Recombinant Protein;Human Reg4 Protein;100UG;Human Reg4, His Tag (RE4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Pro 158 (Accession # AAH17089).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.