Learn More
Cayman Chemical Regadenoson 10mg
SDP
Catalog No. 502980136
Encompass_Preferred
A selective, short-acting adenosine A2A receptor agonist (Ki = 1.1 nM for pig striatum A2A receptor); used to induce hyperemia, particularly in the context of myocardial perfusion imaging
Catalog No. 50-298-0136 Supplier Cayman Chemical Supplier No. 1881410 MG
A selective, short-acting adenosine A2A receptor agonist (Ki = 1.1 nM for pig striatum A2A receptor); used to induce hyperemia, particularly in the context of myocardial perfusion imaging