Learn More
Waters Corp Regenerated Cellulose Membrane, Short Channel, 2 kDa MWCO, Pack of 5
Catalog No. NC2311038 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This regenerated cellulose membrane is designed for use with short channels. It features a 2 kDa molecular weight cut-off and is supplied in a pack of 5, offering compatibility with various accessories.
  • Regenerated cellulose membrane
  • For short channel
  • 2 kDa MWCO
  • Pack of 5
  • Compatible with accessories
Catalog No. NC2311038 Supplier Waters Corp Supplier No. 900326SC02RC2
This regenerated cellulose membrane is designed for use with short channels. It features a 2 kDa molecular weight cut-off and is supplied in a pack of 5, offering compatibility with various accessories.
  • Regenerated cellulose membrane
  • For short channel
  • 2 kDa MWCO
  • Pack of 5
  • Compatible with accessories