Learn More
Medchemexpress LLC Regenerating islet-derived protein 1 beta (REG1B) | >95.0% | 100 UG
SDP
Catalog No. 5000290457
Encompass_Preferred
Recombinant human regenerating islet-derived protein 1 beta (REG1B), expressed in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder. Intended for biochemical and cell-based research applications including studies of tissue regeneration, neuronal growth, and calcium carbonate precipitation; biological activity has been reported in a cell proliferation assay (ED50 ≈ 0.657 μg/mL).
  • Human REG1B protein, full-length mature sequence (Q23-N166).
  • HEK293 expression preserves mammalian post-translational modifications.
  • C-terminal His tag facilitates purification and detection.
  • Lyophilized formulation with low endotoxin (<1 EU/μg) for sensitive assays.
  • High purity (>95.0% by reducing SDS-PAGE) suitable for biochemical work.
  • Biological activity validated in a cell proliferation assay (ED50 ≈ 0.657 μg/mL).
Catalog No. 50-002-90457 Supplier Medchemexpress LLC Supplier No. HYP76565100UG
Recombinant human regenerating islet-derived protein 1 beta (REG1B), expressed in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder. Intended for biochemical and cell-based research applications including studies of tissue regeneration, neuronal growth, and calcium carbonate precipitation; biological activity has been reported in a cell proliferation assay (ED50 ≈ 0.657 μg/mL).
  • Human REG1B protein, full-length mature sequence (Q23-N166).
  • HEK293 expression preserves mammalian post-translational modifications.
  • C-terminal His tag facilitates purification and detection.
  • Lyophilized formulation with low endotoxin (<1 EU/μg) for sensitive assays.
  • High purity (>95.0% by reducing SDS-PAGE) suitable for biochemical work.
  • Biological activity validated in a cell proliferation assay (ED50 ≈ 0.657 μg/mL).