Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Regenerating islet-derived protein 1 beta (REG1B), human recombinant | >95.0% | 10 UG
SDP

Catalog No. 5000289988
Encompass_Preferred

Recombinant human regenerating islet-derived protein 1 beta (REG1B), expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized for use in cell-based assays, biochemical studies, and protein characterization; biological activity is demonstrated in a cell proliferation assay.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Supplied lyophilized from a 0.22 μm filtered PBS solution, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight approximately 19-20 kDa.
  • Biological activity: ED50 0.657 μg/mL in a cell proliferation assay.
  • Recommended storage: lyophilized at -20°C; after reconstitution stable at 4°C for 1 week or -20°C/-80°C for long-term storage.

Catalog No. 50-002-89988 Supplier Medchemexpress LLC Supplier No. HYP7656510UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human regenerating islet-derived protein 1 beta (REG1B), expressed in HEK293 cells with a C-terminal His tag. Supplied lyophilized for use in cell-based assays, biochemical studies, and protein characterization; biological activity is demonstrated in a cell proliferation assay.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • Supplied lyophilized from a 0.22 μm filtered PBS solution, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight approximately 19-20 kDa.
  • Biological activity: ED50 0.657 μg/mL in a cell proliferation assay.
  • Recommended storage: lyophilized at -20°C; after reconstitution stable at 4°C for 1 week or -20°C/-80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.