Learn More
MOLTOX Molecular Toxicology Inc Lyophilized NADPH REGENSYS B, 31.5 mg of crystalline NADP, Add to Regensys A. Pkg w/ dessicant, Store in freezer
SDP
Catalog No. NC1088347
Encompass
Convenient preformulated filter sterilized and prepackaged S9 mix components for preparation of 4 - 10% S9 mix. Regensys™ "A" is formulated as described by Maron & Ames, Mut. Res. 113: 173, 1983. Size refers to final volume of S9 mix. Regensys™ "B" consists of accurately weighed aliquots of crystalline NADP to add to corresponding Regensys™ "A" for completion of the Regensys™ system. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide.
Catalog No. NC1088347 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 60201.3L
Convenient preformulated filter sterilized and prepackaged S9 mix components for preparation of 4 - 10% S9 mix. Regensys™ "A" is formulated as described by Maron & Ames, Mut. Res. 113: 173, 1983. Size refers to final volume of S9 mix. Regensys™ "B" consists of accurately weighed aliquots of crystalline NADP to add to corresponding Regensys™ "A" for completion of the Regensys™ system. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide.