Learn More
Regin Hbac Products Smoke Pen
Catalog No. NC9432464
Encompass_Preferred
The Regin Smoke Pen is truly an innovation in the world of smoke testing. This innovative product works like a mechanical pencil. Our patented smoke emitting wick is inserted into the convenient to use pen, adjust the wick to provide 3/8″ of exposed wick, light the wick with a match or lighter. The Smoke Pen will provide a continuous trail of smoke. The Smoke Pen can then be easily extinguished by simply replacing the protective cover. The Smoke Pen can then be stored and re-used for the next test. This non-toxic, cool burning emitter is ideal for air balancing, verifying ventilation, testing smoke alarms, testing for negative or positive pressure and more.
Catalog No. NC9432464 Supplier Regin Hbac Products Supplier No. S220 SMOKE PEN
The Regin Smoke Pen is truly an innovation in the world of smoke testing. This innovative product works like a mechanical pencil. Our patented smoke emitting wick is inserted into the convenient to use pen, adjust the wick to provide 3/8″ of exposed wick, light the wick with a match or lighter. The Smoke Pen will provide a continuous trail of smoke. The Smoke Pen can then be easily extinguished by simply replacing the protective cover. The Smoke Pen can then be stored and re-used for the next test. This non-toxic, cool burning emitter is ideal for air balancing, verifying ventilation, testing smoke alarms, testing for negative or positive pressure and more.