Learn More
Regis Technologies Inc GC Regeant, Regis, REGISIL (BSTFA) + 1% TMCS (25 GM)
SDP
Supplier:  Regis Technologies Inc 1270123200
Encompass
BSTFA +1% TMCS-Regisil is a Silylation Reagent. BSTFA reacts faster and more completely than BSA, due to the trifluoroacetamide leaving group. The high volatility of BSTFA and its byproducts results in non-co-elution of early eluting peaks. Additionally, the highly volatile and stable products result in low detector noise and fouling. Addition of TMCS to BSTFA catalyzes reactions of hindered functional groups and other difficult functionalities, such as secondary hydroxyls and amines.
Catalog No. 50-442-884