Learn More
Regis Technologies Inc GC Regeant, Regis, REGISIL (BSTFA) (25 GM)
SDP
Supplier:  Regis Technologies Inc 1270113200
Encompass
BSTFA-Regisil is a Silylation Reagent. BSTFA reacts faster and more completely than BSA, due to the trifluoroacetamide leaving group. The high volatility of BSTFA and its byproducts results in non-co-elution of early eluting peaks. Additionally, the highly volatile and stable products result in low detector noise and fouling.
Catalog No. 50-442-880