Learn More
Ismatec™ Reglo Miniflex Dispensing Pump Systems
Catalog No. 11475091
Change view
Click to view available options
Model:
System with Remote Control
Catalog No. Model
11-475-091 System with Remote Control
1 options
Catalog No. 11-475-091 Supplier Ismatec™ Supplier No. MFLX7792222
Add to Cart
Add to Cart
Request a Quote
Complete systems to increase the efficiency and accuracy of filling and dispensing applications
  • Flow range 0.16 to 365 mL/min with included L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Realize increased efficiency and throughput when compared to hand pouring, manually pipetting, or using bottle-top style dispensers
  • 5 in. (12.7 cm) capacitive TFT/RGB touch screen puts the intuitive interface at your fingertips
  • Dispense by volume or time -- anti-drip and speed ramping available in Volume dispense mode
  • Flexible operational control via screen, MasterflexLive, or analog remote control
  • Pump lets you save and retrieve commonly used dispense protocols for rapid set up
Specifications
Control Type Digital Variable Speed
Display TFT Touchscreen
Electrical Requirements 90 to 260 V, 50/60 Hz
Flow Rate 0.16 to 365 mL/min.
IP Rating IP31
Includes PTFE Tube Weight, Tubing (3 x 8 ft.)
Model System with Remote Control
Reversible Yes
Maximum Pressure Dependent on tubing