Learn More
Medchemexpress LLC HY-10331 100mg Medchemexpress, Regorafenib CAS:755037-03-7 Purity:>98%
SDP
Catalog No. 502028727
Encompass_Preferred
Medchemexpress, HY-10331 100mg Regorafenib CAS:755037-03-7 Regorafenib (BAY 73-4506) is a multi-targeted receptor tyrosine kinase inhibitor with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8727 Supplier Medchemexpress LLC Supplier No. HY10331100MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-10331 100mg Regorafenib CAS:755037-03-7 Regorafenib (BAY 73-4506) is a multi-targeted receptor tyrosine kinase inhibitor with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.