Learn More
Cayman Chemical RegorafenIb-13C-d3 500ug
SDP
Catalog No. 503015827
Encompass_Preferred
An internal standard for the quantification of regorafenib by GC- or LC-MS
Catalog No. 50-301-5827 Supplier Cayman Chemical Supplier No. 25486500 UG
An internal standard for the quantification of regorafenib by GC- or LC-MS