Learn More
Apexbio Technology LLC Regorafenib 755037-03-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062185
Encompass_Preferred
Regorafenib (CAS 755037-03-7) is an orally bioavailable inhibitor targeting multiple receptor tyrosine kinases (RTKs) such as VEGFR1/2/3 PDGFR Kit RET Raf-1 B-RAF and B-RAFV600E By impeding kinase phosphorylation (IC50 values ranging from approximately 1 5 46 nM) regorafenib disrupts signaling pathways associated with angiogenesis tumor cell proliferation and metastasis Preclinically regorafenib inhibited VEGFR2 phosphorylation (IC50 3 nM) and reduced VEGF-induced endothelial cell proliferation in vitro In rodent tumor xenograft models it demonstrated dose-dependent tumor growth suppression and antimetastatic potential supporting its utility for oncology research
Catalog No. 50-000-62185 Supplier Apexbio Technology LLC Supplier No. A82365.1
Regorafenib (CAS 755037-03-7) is an orally bioavailable inhibitor targeting multiple receptor tyrosine kinases (RTKs) such as VEGFR1/2/3 PDGFR Kit RET Raf-1 B-RAF and B-RAFV600E By impeding kinase phosphorylation (IC50 values ranging from approximately 1 5 46 nM) regorafenib disrupts signaling pathways associated with angiogenesis tumor cell proliferation and metastasis Preclinically regorafenib inhibited VEGFR2 phosphorylation (IC50 3 nM) and reduced VEGF-induced endothelial cell proliferation in vitro In rodent tumor xenograft models it demonstrated dose-dependent tumor growth suppression and antimetastatic potential supporting its utility for oncology research