Learn More
AdipoGen Regorafenib (5 mg)
SDP
Supplier:  AdipoGen AGCR13623M005
Encompass_Preferred
Chemical. CAS: 755037-03-7. Formula: C21H15ClF4N4O3. MW: 482.8. Orally bioavailable potent antitumor, antiangiogenic, antiproliferative and antineoplastic agent. Multi-target kinase inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRbeta, Kit, RET and Raf-1 with IC50 values of 13, 4.2, 46, 22, 7, 1.5 and 2.5nM, respectively.
Catalog No. 50-205-3993