Learn More
AdipoGen Regorafenib BAY 73-4506
SDP
Supplier:  AdipoGen AGCR13623M025
Encompass_Preferred
Chemical. CAS 755037-03-7. Formula C21H15ClF4N4O3. MW 482.8. Orally bioavailable potent antitumor, antiangiogenic, antiproliferative and antineoplastic agent. Multi-target kinase inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRbeta, Kit, RET and Raf-1 with IC50 values of 13, 4.2, 46, 22, 7, 1.5 and 2.5nM, respectively.
Catalog No. 50-256-2263