Learn More
Apexbio Technology LLC Regorafenib hydrochloride(Synonyms: Regorafenib HCl, BAY 73-4506, Stivarga, BAY73-4506 hydrochloride, Regorafenib hydrochloride salt), 10mM (in 1mL DMSO), CAS: 835621-07-3.
SDP
Catalog No. 5000060309
Encompass_Preferred
Regorafenib hydrochloride (CAS 835621-07-3) is an orally bioavailable small molecule inhibitor targeting multiple protein kinases including c-KIT VEGFR2 and B-Raf It inhibits these kinases with IC50 values of approximately 17 nM (c-KIT) 40 nM (VEGFR2) and 69 nM (B-Raf) By blocking kinase signaling Regorafenib hydrochloride effectively interrupts tumor-associated pathways involved in cell proliferation angiogenesis and survival It is widely used in oncology research for studies focused on kinase-mediated tumor growth and vascularization mechanisms
Catalog No. 50-000-60309 Supplier Apexbio Technology LLC Supplier No. A37505.1
Regorafenib hydrochloride (CAS 835621-07-3) is an orally bioavailable small molecule inhibitor targeting multiple protein kinases including c-KIT VEGFR2 and B-Raf It inhibits these kinases with IC50 values of approximately 17 nM (c-KIT) 40 nM (VEGFR2) and 69 nM (B-Raf) By blocking kinase signaling Regorafenib hydrochloride effectively interrupts tumor-associated pathways involved in cell proliferation angiogenesis and survival It is widely used in oncology research for studies focused on kinase-mediated tumor growth and vascularization mechanisms