Learn More
TARGETMOL CHEMICALS INC REGORAFENIB HYDROCHLORID 100MG
SDP
Catalog No. 502579080
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Regorafenib Hydrochloride (BAY73-4506 hydrochloride) is a new oral multikinase inhibitor of angiogenic stromal and oncogenic receptor tyrosine kinases with potent preclinical antitumor activitypurity: 100%
Catalog No. 50-257-9080 Supplier TARGETMOL CHEMICALS INC Supplier No. T8402100MG
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Regorafenib Hydrochloride (BAY73-4506 hydrochloride) is a new oral multikinase inhibitor of angiogenic stromal and oncogenic receptor tyrosine kinases with potent preclinical antitumor activitypurity: 100%