Learn More
Selleck Chemical LLC Regorafenib Hydrochloride-25mg
SDP
Catalog No. 502389665
Encompass_Preferred
Regorafenib (Stivarga, BAY 73-4506) Hydrochloride is a multi-target inhibitor for VEGFR1, Murine VEGFR2/3, PDGFRβ, Kit (c-Kit), RET (c-RET) and Raf-1 with IC50 of 13 nM, 4.2 nM/46 nM, 22 nM, 7 nM, 1.5 nM and 2.5 nM, respectively.
Catalog No. 50-238-9665 Supplier Selleck Chemical LLC Supplier No. S494725MG
Regorafenib (Stivarga, BAY 73-4506) Hydrochloride is a multi-target inhibitor for VEGFR1, Murine VEGFR2/3, PDGFRβ, Kit (c-Kit), RET (c-RET) and Raf-1 with IC50 of 13 nM, 4.2 nM/46 nM, 22 nM, 7 nM, 1.5 nM and 2.5 nM, respectively.