Learn More
Medchemexpress LLC Regorafenib hydrochloride | 835621-07-3 | MFCD22417053 | C21H16Cl2F4N4O3 | 10MG
SDP
Catalog No. 5000260715
Encompass_Preferred
Regorafenib hydrochloride is the hydrochloride salt of regorafenib, a potent multi-target kinase inhibitor. Supplied as an analytical standard for research use, it inhibits VEGFR1/2/3, PDGFRβ, Kit, RET, and Raf-1 with low-nanomolar potency, and is intended for in vitro and analytical applications such as HPLC, GC, and MS.
  • Multi-target kinase inhibitor active against VEGFR1/2/3, PDGFRβ, Kit, RET, and Raf-1.
  • Low-nanomolar potency reported for key kinase targets.
  • High purity analytical standard (99.6%).
  • Intended for research and analytical applications, including HPLC, GC, and MS.
  • Suitable for method development and quantitative assays.
Catalog No. 50-002-60715 Supplier Medchemexpress LLC Supplier No. HY1330810MG
Regorafenib hydrochloride is the hydrochloride salt of regorafenib, a potent multi-target kinase inhibitor. Supplied as an analytical standard for research use, it inhibits VEGFR1/2/3, PDGFRβ, Kit, RET, and Raf-1 with low-nanomolar potency, and is intended for in vitro and analytical applications such as HPLC, GC, and MS.
  • Multi-target kinase inhibitor active against VEGFR1/2/3, PDGFRβ, Kit, RET, and Raf-1.
  • Low-nanomolar potency reported for key kinase targets.
  • High purity analytical standard (99.6%).
  • Intended for research and analytical applications, including HPLC, GC, and MS.
  • Suitable for method development and quantitative assays.