Learn More
Medchemexpress LLC HY-10331A 100mg Medchemexpress, Regorafenib (monohydrate) CAS:1019206-88-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10331A100MG
Encompass_Preferred
Medchemexpress, HY-10331A 100mg Regorafenib (monohydrate) CAS:1019206-88-2 Regorafenib monohydrate is a multi-target inhibitor for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1 with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8075