Learn More
Apexbio Technology LLC Regorafenib monohydrate 1019206-88-2 100mg
SDP
Catalog No. 5000063997
Encompass_Preferred
Regorafenib monohydrate (CAS 1019206-88-2) is a multitargeted tyrosine kinase inhibitor that suppresses numerous intracellular and membrane receptor tyrosine kinases (RTKs) including VEGFR (types 1-3) PDGFR- FGFR-1 RET TIE2 c-KIT Raf-1 and BRAF (wild-type and V600E mutant) It inhibits angiogenic RTKs such as VEGFR-2 with IC50 values ranging from approximately 4 2 to 46 nM and oncogenic RTKs like c-KIT and RET with IC50 values around 1 5 to 28 nM Regorafenib demonstrates antiproliferative activity in vitro (mean IC50 1 M) and reduces tumor progression in xenograft animal models supporting its use in cancer research involving breast colorectal thyroid pancreatic cancers melanoma and gastrointestinal stromal tumors
Catalog No. 50-000-63997 Supplier Apexbio Technology LLC Supplier No. A3751100
Regorafenib monohydrate (CAS 1019206-88-2) is a multitargeted tyrosine kinase inhibitor that suppresses numerous intracellular and membrane receptor tyrosine kinases (RTKs) including VEGFR (types 1-3) PDGFR- FGFR-1 RET TIE2 c-KIT Raf-1 and BRAF (wild-type and V600E mutant) It inhibits angiogenic RTKs such as VEGFR-2 with IC50 values ranging from approximately 4 2 to 46 nM and oncogenic RTKs like c-KIT and RET with IC50 values around 1 5 to 28 nM Regorafenib demonstrates antiproliferative activity in vitro (mean IC50 1 M) and reduces tumor progression in xenograft animal models supporting its use in cancer research involving breast colorectal thyroid pancreatic cancers melanoma and gastrointestinal stromal tumors