Learn More
Medchemexpress LLC Regorafenib monohydrate | 1019206-88-2 | MFCD17170366 | 99.7% | 500.8 g/mol | C21H17ClF4N4O4 | 1 ML
SDP
Catalog No. 5000247539
Encompass_Preferred
Regorafenib monohydrate is the monohydrate form of regorafenib, a multi-targeted receptor tyrosine kinase inhibitor used in research. The item is provided as a 10 mM solution in a 1 mL vial for laboratory and analytical applications; it is intended for research use only.
  • High purity suitable for research (purity 99.7%).
  • Monohydrate form for consistent chemical properties.
  • Concentrated solution (10 mM) enabling small-volume dosing.
  • Convenient 1 mL vial for storage and handling.
  • Useful as a kinase inhibitor in in vitro studies.
Catalog No. 50-002-47539 Supplier Medchemexpress LLC Supplier No. HY10331A10MM 1ML
Regorafenib monohydrate is the monohydrate form of regorafenib, a multi-targeted receptor tyrosine kinase inhibitor used in research. The item is provided as a 10 mM solution in a 1 mL vial for laboratory and analytical applications; it is intended for research use only.
  • High purity suitable for research (purity 99.7%).
  • Monohydrate form for consistent chemical properties.
  • Concentrated solution (10 mM) enabling small-volume dosing.
  • Convenient 1 mL vial for storage and handling.
  • Useful as a kinase inhibitor in in vitro studies.