Learn More
Medchemexpress LLC Regorafenib monohydrate | 1019206-88-2 | MFCD17170366 | 99.9% | 500.8 g/mol | C21H17ClF4N4O4 | 50 MG
SDP
Catalog No. 5000244934
Encompass_Preferred
Regorafenib monohydrate is the monohydrate form of regorafenib, a multi-targeted receptor tyrosine kinase inhibitor used as a research reagent and analytical standard. It exhibits potent inhibition of VEGFR, PDGFR, KIT, RET, and RAF kinases and is used in preclinical assays to study antitumor and antiangiogenic activity.
  • High purity (≈99.9%) suitable for analytical work.
  • Monohydrate form with molecular weight 500.8 g/mol.
  • Useful as an analytical standard and research reagent in kinase assays.
  • Available in small, quantified laboratory packages for preclinical studies.
Catalog No. 50-002-44934 Supplier Medchemexpress LLC Supplier No. HY10331A50MG
Regorafenib monohydrate is the monohydrate form of regorafenib, a multi-targeted receptor tyrosine kinase inhibitor used as a research reagent and analytical standard. It exhibits potent inhibition of VEGFR, PDGFR, KIT, RET, and RAF kinases and is used in preclinical assays to study antitumor and antiangiogenic activity.
  • High purity (≈99.9%) suitable for analytical work.
  • Monohydrate form with molecular weight 500.8 g/mol.
  • Useful as an analytical standard and research reagent in kinase assays.
  • Available in small, quantified laboratory packages for preclinical studies.