Learn More
TARGETMOL CHEMICALS INC REGRELOR DISODIUM 5MG
SDP
Catalog No. 502580271
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Regrelor disodium (INS 50589) is a p2y12 receptor antagonist that prevents thrombosis effectively inhibits vascular smooth muscle cell contraction inhibits cell proliferation and reduces inflammation. purity: 98%
Catalog No. 50-258-0271 Supplier TARGETMOL CHEMICALS INC Supplier No. T342805MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Regrelor disodium (INS 50589) is a p2y12 receptor antagonist that prevents thrombosis effectively inhibits vascular smooth muscle cell contraction inhibits cell proliferation and reduces inflammation. purity: 98%