Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Regular Grade Leather Drivers Gloves

Catalog No. 19071377 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-071-377 X-Large
19-071-371 Small
19-071-373 Medium
19-071-375 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-071-377
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves provide abrasion resistance and dexterity

  • Ideal for construction, machine operation, utility work, farming, roping, and other general applications
  • Top-grain leather
  • Keystone thumb
  • Launderable

Specifications

Color Natural
Grade Regular
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size X-Large
Style Standard
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, Roping, and Many Other General Applications
Glove Cuff Style Open
Hem Color Blue
Thumb Style Keystone
Length (English) 10.3 in.
Length (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.