Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Regular Grade Top Grain Cowhide Leather Palm Gloves with Fabric Back

Catalog No. 19048666A Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-666A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-048-666A
  • $222.00
    $133.65 / Dozen of 12

    Save $88.35

    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,158.00
    $697.65 / Case of 6 DZ

    Save $460.35

    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cowhide leather construction provides comfort, durability, abrasion resistance, and breathability. Fabric back ensures breathability and flexibility.

  • Fabric back provides breathability and flexibility
  • Leather knuckle strap and finger tips for extra protection
  • Cotton lined palm for extra comfort
  • Elastic closure sewn inside glove for safety and comfort
  • Gauntlet cuff for added forearm protection and allows for easy on and off
  • Gunn cut pattern delivers durability, wear, and comfort
  • Wing thumb increases comfort in open-handed work
  • Color coded hems for easy size identification

Specifications

Back Material Fabric
Cuff Material Rubberized
Color Natural
Grade Regular
Hand Specific
Material Top Grain Cowhide Leather
Product Type Leather Palm Gloves
Size Small
Style Gunn
Glove Cuff Color Blue with Red Stripes
Glove Cuff Style Gauntlet
Hem Color Red
Liner Material Cotton
Series 87-1663
Thumb Style Wing
Length (English) 11.3 in.
Length (English) Cuff 4.5 in.
Length (Metric) 28.8 cm
Length (Metric) Cuff 11.5 cm
Show More Show Less

Warning - Prop 65 - Cancer and Reproductive Harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.