Learn More
Sklar Instruments Econo Regular Splinter Forceps, Sklar, 3-1/2 in., Sharp, Straight, Serrated, Stainless Steel. PK/12.
SDP
Catalog No. 501437684 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Regular Splinter Forceps, Sklar, 3-1/2 in., Sharp, Straight, Serrated, Stainless Steel. Econo Splinter Forceps are multi-purpose thumb forceps with sharp tips used for removing small foreign objects. Econo instruments are Sklar's floor-grade, disposable instruments. Made in Pakistan, they offer the greatest utility at the lowest cost. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 3-1/2 inches. It is sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7684 Supplier Sklar Instruments Supplier No. 21777
Econo Regular Splinter Forceps, Sklar, 3-1/2 in., Sharp, Straight, Serrated, Stainless Steel. Econo Splinter Forceps are multi-purpose thumb forceps with sharp tips used for removing small foreign objects. Econo instruments are Sklar's floor-grade, disposable instruments. Made in Pakistan, they offer the greatest utility at the lowest cost. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 3-1/2 inches. It is sold in boxes of 12.