Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plas Labs™ Regular Stream Table
SDPSCI_ED

Catalog No. S68802 Shop All Plas-Labs, Inc. Products
Change view
Click to view available options
Plas Lab™ Stream Table Regular
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S68802 Plas Lab™ Stream Table Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S68802 Supplier Plas Labs™ Supplier No. 105RT
This item is not returnable. View return policy

Designed and created to facilitate the simulations and study of many basic land forms for Earth Science.

Designed and created to facilitate the simulations and study of many basic land forms for Earth Science.

  • Includes wave maker with motor, shaft and paddles, power pack, water pump, and hose.
  • 60' long x 20' wide x 6' deep

Specifications

Grade 6+
For Use With (Application) Facilitates the simulations and study of many basic land forms for Earth Science
Product Type Teaching Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.