Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Regular Weight Brown Jersey Gloves

Catalog No. 1139376 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
11-393-76 Men's
11-462-1 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-393-76 Supplier Wells Lamont™ Supplier No. Y7201L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Regular Weight Brown Jersey Gloves are constructed from a combination of polyester/cotton, providing comfort and flexibility while on the job. Knit wrist helps prevent irritants from entering the glove.

  • Poly/cotton blend offers increased pliability and elasticity
  • Dark color helps conceal dirt, extending the wear life of the glove
  • Knit wrist helps keep debris out
  • Clute cut

Specifications

Cleaning Procedure Machine Washable
Construction Jersey Knit
Applications Industrial
Coating Material Uncoated
Color Brown
Disposable Reusable
Exterior Finish No Texture
Hand Specific
Material Polycotton
Product Type General Purpose Gloves
Size Large
Sterility Non-sterile
Style Clute
For Use With (Application) Construction, Warehousing
Gender Men's
Glove Cuff Color Brown
Glove Cuff Style Knit
Grip Design Smooth
Lining Unlined
Packaging Type Bulk
Palm Type Standard
Quantity 12 Pairs
Shell Material Poly/Cotton
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.