Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Toter Hazardous Waste Can, 64 gal., Rectangular, Red, 1 ea

Catalog No. 502816463
Encompass

This hazardous waste can is designed for the safe collection and containment of hazardous materials. It features a robust design suitable for industrial and laboratory environments.

  • Container capacity: 64 gal.
  • Rectangular shape
  • Overall diameter: 24.25 in.
  • Overall height: 41.75 in.
  • Red color
  • Country of origin: Mexico

Catalog No. 50-281-6463 Supplier MSC Supplier No. 17929308
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This hazardous waste can is designed for the safe collection and containment of hazardous materials. It features a robust design suitable for industrial and laboratory environments.

  • Container capacity: 64 gal.
  • Rectangular shape
  • Overall diameter: 24.25 in.
  • Overall height: 41.75 in.
  • Red color
  • Country of origin: Mexico

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.