Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sterilis Solutions Regulated Medical Waste (RMW) Autoclave/Grinding System
SDP

Catalog No. 23777000
Change view
Click to view available options
Capacity (Metric):
19 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23-777-000 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 23-777-000 Supplier Sterilis Solutions Supplier No. 38233000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Sterilis Solutions™ Regulated Medical Waste (RMW) Autoclave/Grinding System provides on-demand remediation of RMW at the point of use.

  • A new, revolutionary approach to medical waste that combines an autoclave and grinder solution
  • Takes sharps containers and red bag waste from biohazardous waste (RMW) to harmless right at the point of use
  • The inert, confetti-like material that remains can be thrown out with the regular, urban waste stream.
  • Safety increased & Liability reduced
    • Point-of-care solution remediates RMW on demand, reducing the number of touch points and eliminating hauling liability
  • Reduces labor and handling
    • Easy-to-use device and touch pad
    • Cloud-based recording of  data
    • No manual tracking or record-keeping
    • No aggregating or boxing or shipping preparation
  • Dramatically improves sustainability
    • Reduces carbon footprint (CO2, mercury emissions, and incinerator ash) with waste volume reduction up to 80%
  • Significant cost savings
    • Provides controllable fixed costs
    • Labor/regulatory cost savings
    • Lower operating costs
  • Fortifies emergency preparedness
    • Portable, self-contained system
    • Plug-in and start
  • Versatility
  • Designed to handle small to medium amounts of waste
  • The size makes it an excellent choice for clinics, labs, surgery centers, quarantine areas, dialysis, dermatology, and ophthalmology practices
  • Easily fits into a clinic, lab, or dirty room
  • User Interface Touch Screen; Log in, load RMW and press start
  • Approximately 60-minute cycle
  • Automatically logs and stores compliance data
  • Prompts and records  biochallenges to state regulations
  • Low operating noise, < 60dB
  • No plumbing required, self contained system

Specifications

Amperage 30 A
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Depth (Metric) Chamber 355 mm
Diameter (English) Chamber 13 in.
Depth (English) Chamber 14 in.
Diameter (Metric) Chamber 330 mm
For Use With (Application) Biohazardous RMW
Length (English) 36 in.
Height (English) Exterior 48 in.
Height (Metric) Exterior 1194 mm
Length (Metric) 914 mm
Model 3000
Product Type Autoclave/Grinder
Temperature (English) 269°F to 280°F
Temperature (Metric) 132°C to 138°C
Weight (English) 936 lb.
Display Touchscreen
Weight (Metric) 425 kg
Electrical Requirements Single Phase
Frequency 60 Hz
Interface User Friendly
Pressure (English) Air Supply Self Contained
Pressure (Metric) Feed Water Self Contained
Pressure (English) Feed Water Self Contained
Pressure (Metric) Air Supply Self Contained
Sterilization Time 60 min.
Voltage 220 V
Width (English) Exterior 26 in.
Sterility Sterile
Width (Metric) Exterior 607 mm
Dimensions (L x W x H) 36 x 26 x 48 in.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.