Learn More
Hach Company Pressure Regulator, Preset 4 psi for 5500sc Analyzers
Catalog No. NC2590875 Shop All Hach Company Products
Encompass
Pressure regulator preset to 4 psi (0.28 bar) designed for use with 5500sc series analyzers. It limits sample-line pressure to protect the analyzer from damage and helps ensure stable sample delivery for reliable measurements.
  • Preset pressure to 4 psi (0.28 bar).
  • Protects analyzer from higher sample-line pressure.
  • Compatible with 5500sc series analyzers.
  • Helps maintain stable sample flow for reliable results.
Catalog No. NC2590875 Supplier Hach Company Supplier No. 6782900
Pressure regulator preset to 4 psi (0.28 bar) designed for use with 5500sc series analyzers. It limits sample-line pressure to protect the analyzer from damage and helps ensure stable sample delivery for reliable measurements.
  • Preset pressure to 4 psi (0.28 bar).
  • Protects analyzer from higher sample-line pressure.
  • Compatible with 5500sc series analyzers.
  • Helps maintain stable sample flow for reliable results.