Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rehmannic acid | 467-81-2 | 552.78 | 1 MG
SDP

Catalog No. 5000415633
Encompass_Preferred

Rehmannic acid (lantadene A) is a compound isolated from Lantana camara. It demonstrates considerable in vitro antioxidant and free radical scavenging activities in a dose-dependent manner. Rehmannic acid is considered a promising candidate for use as an antioxidant and hepatoprotective agent.

  • Considerable in vitro antioxidant activity
  • Free radical scavenging capacity activities
  • Promising candidate for use as an antioxidant and hepatoprotective agent
  • Isolated from Lantana camara
  • Purity: 96.0%
  • Appearance: Solid, white to off-white

Catalog No. 50-004-15633 Supplier Medchemexpress LLC Supplier No. HYN15211MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rehmannic acid (lantadene A) is a compound isolated from Lantana camara. It demonstrates considerable in vitro antioxidant and free radical scavenging activities in a dose-dependent manner. Rehmannic acid is considered a promising candidate for use as an antioxidant and hepatoprotective agent.

  • Considerable in vitro antioxidant activity
  • Free radical scavenging capacity activities
  • Promising candidate for use as an antioxidant and hepatoprotective agent
  • Isolated from Lantana camara
  • Purity: 96.0%
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.