Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rehmannioside C | 81720-07-2 | 5 MG
SDP

Catalog No. 5000421421
Encompass_Preferred

Rehmannioside C is an iridoid glucoside isolated from Radix Rehmanniae Praeparata.

  • Purity of 99.47%
  • Molecular formula C21H34O14
  • Molecular weight of 510.49
  • Appears as a solid
  • Color from white to light yellow
  • Soluble in DMSO
  • Standard storage at 4°C, sealed, away from moisture and light

Catalog No. 50-004-21421 Supplier Medchemexpress LLC Supplier No. HYN24005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rehmannioside C is an iridoid glucoside isolated from Radix Rehmanniae Praeparata.

  • Purity of 99.47%
  • Molecular formula C21H34O14
  • Molecular weight of 510.49
  • Appears as a solid
  • Color from white to light yellow
  • Soluble in DMSO
  • Standard storage at 4°C, sealed, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.