Learn More
TARGETMOL CHEMICALS INC Rehmannioside D 10MG
SDP
Catalog No. 502879975
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Rehmannioside D is a carotenoid glycoside that existed in the roots of Rehmannia glutinosa. Purity 98.67%
Catalog No. 50-287-9975 Supplier TARGETMOL CHEMICALS INC Supplier No. T702510MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Rehmannioside D is a carotenoid glycoside that existed in the roots of Rehmannia glutinosa. Purity 98.67%