Learn More
Bellco Glass, Inc. Reich Slide,25x75mm
SDP
Catalog No. 501945823
Encompass_Preferred
Modification of the Breed Counting Technique. The slide spreads a known volume of solution over a known area and permits a calculation of bacteria per sample. It has 3 rings with each interior area measuring 1cm squared. MADE IN THE USA Measurement: 25x75cm
Catalog No. 50-194-5823 Supplier Bellco Glass, Inc. Supplier No. 563801920
Modification of the Breed Counting Technique. The slide spreads a known volume of solution over a known area and permits a calculation of bacteria per sample. It has 3 rings with each interior area measuring 1cm squared. MADE IN THE USA Measurement: 25x75cm