Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reichert™ Clinic-Chek Digital Handheld Refractometer

Catalog No. 13940021 Shop All Reichert, Inc Products
Change view
Click to view available options
Scale Range:
1 to 1.04 Urine Specific Gravity, 0 to 15 g/100 mL Serum Protein
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scale Range
13-940-021 1 to 1.04 Urine Specific Gravity, 0 to 15 g/100 mL Serum Protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-940-021 Supplier Reichert™ Supplier No. 13940021
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

2 AAA batteries

Suitable for medical clinic, laboratory and athletic/wrestling hydration measurement

  • Programmed with three measurement scales (urine specific gravity, serum protein concentration, refractive index)
  • One unit required for measurement
  • IP65 dustproof/water resistant rating
  • More accurate than hydrometer and test strips
  • 5 digit full view LCD screen displays all pertinent measurement data in one view
  • Easy to operate with reading errors eliminated
  • With automatic shutdown feature

  • No need to place sample near face while measurement
  • Safe and convenient measurement
  • For over 10000 measurements
  • Provides quick results
  • Prism material: Glass

For measuring body fluids (urine and blood serum)

Certifications

CE marked

Compliance

RoHS, WEEE

Warranty and Services

One year

Specifications

Type Handheld Clinic-Chek Digital Refractometer
Scale Range 1 to 1.04 Urine Specific Gravity, 0 to 15 g/100 mL Serum Protein
Refractive Index 1.333 to 1.4465
Calibration Water
Accuracy 0.005 USG, 0.1 Serum Protein
For Use With (Application) For measuring body fluids (Urine and Blood Serum)
Includes 2 AAA Batteries
Certifications/Compliance IP65, RoHS, WEEE, CE
Weight (English) 3.5 oz.
Weight (Metric) 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.