Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reichert™ TS Meter-D Automatic Digital Clinical Refractometer

Catalog No. 1360000 Shop All Reichert, Inc Products
Change view
Click to view available options
TS Meter-D Automatic Digital Clinical Refractometer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-600-00 TS Meter-D Automatic Digital Clinical Refractometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-600-00 Supplier Reichert™ Supplier No. 13960000

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Automatically read samples with precision comparable to high-end automatic refractometers.

  • New sapphire prism for greater durability and accuracy
  • New sapphire prism delivers 2.5x more conductivity to temperature, providing greater accuracy of the Automatic Temperature Compensation (ATC) calculation
  • Wide reading range enables use of just one instrument to measure different samples
  • Direct reading of urine specific gravity, copper sulfate specific gravity, serum or plasma protein concentration all with great precision
  • Touch-of-a-button easy
  • No more guessing about fuzzy shadowlines
  • PC connectivity for transfer of measurements and programming of scales
  • IR Communications Package enables reading log, calibration log, and custom scale transfer to any lab PC

Specifications

  • Reading Range: 1.000 to 1.060 Urine Specific Gravity; 0.0 to 15.0g/100mL Serum Protein; 1.000 to 1.200 Copper Sulfate Specific Gravity; 1.3300 to 1.5600 Refractive Index
  • Scale: Refractive Index, Urine Specific Gravity, Serum Protein, Copper Sulfate
  • Precision: 0.0001 Refractive Index; ±0.001 Urine Specific Gravity; ±0.001 Copper Specific Gravity; ±0.1 g/100mL Serum Protein
  • Accuracy: ±0.0001
  • Calibration: Set point or set point/span user definable
  • Prism Assembly: Sapphire in stainless steel well
  • I/O: IR Docking Station
  • Power: 4-AAA alkaline batteries (included)
  • Dimensions (L x W x H): 18 x 9 x 3.5cm

Warranty and Services

Two years

Specifications

Type Automatic Digital Clinical Refractometer
Scale Range 1.000 to 1.060 Urine Specific Gravity, 0.0 to 15.0 g/100 mL Serum Protein, 1.000 to 1.200 Copper Sulfate Specific Gravity
Refractive Index 1.3300 to 1.5600
Model TS Meter-D
Calibration Set point or set point/span user definable
Accuracy ±0.0001
For Use With (Application) Direct reading of urine specific gravity, copper sulfate specific gravity, serum or plasma protein concentration
Certifications/Compliance NIST
Dimensions (L x W x H) 18 x 9 x 3.5 cm
Power Supply Details 4xAAA Alkaline Batteries
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.