Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reichert™ USG-Chek Digital Handheld Refractometer

Catalog No. 13940019 Shop All Reichert, Inc Products
Change view
Click to view available options
Scale Range:
1 to 1.04
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scale Range
13-940-019 1 to 1.04
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-940-019 Supplier Reichert™ Supplier No. 13940019
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

2 AAA batteries

Urine specific gravity refractometer suited for medical clinic

  • Delivers fast and accurate hydration measurement in athletics, particularly wrestling
  • More accurate than hydrometers and test strips
  • Easy to operate and provides quick results
  • 5-digit full view LCD screen displays all pertinent measurement data in one view
  • For over 10000 measurements with no reading errors
  • Has automatic shutdown feature that preserves battery life
  • Ratings: IP65 Dustproof / Water Resistant

  • No need to place sample near face while measuring
  • Safe and convenient measurement
  • Prism material: Glass

Ideal for use with sports hydration: wrestling, track and field and team sports

Certifications

CE marked

Compliance

RoHS, WEEE

Warranty and Services

One Year

Specifications

Scale Range 1 to 1.04
Calibration Water
Accuracy 0.005
Includes 2 AAA Batteries
Certifications/Compliance IP65, RoHS, WEEE, CE
Weight (English) 3.5 oz.
Weight (Metric) 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.