Learn More
Waters Corp Manifold Assembly, REIMS, Mechanical, MS System
Catalog No. 502391473 Shop All Waters Corporation Products
Encompass_Preferred
This replacement manifold assembly is specifically designed for the REIMS Source. It is a mechanical component suitable for MS Systems, and comes complete with necessary fittings and a restrictor.
  • Specifically designed for the REIMS Source
  • Mechanical type
  • Suitable for MS systems
  • Includes fittings and restrictor
  • Replacement as required
Catalog No. 50-239-1473 Supplier Waters Corp Supplier No. 700011620
This replacement manifold assembly is specifically designed for the REIMS Source. It is a mechanical component suitable for MS Systems, and comes complete with necessary fittings and a restrictor.
  • Specifically designed for the REIMS Source
  • Mechanical type
  • Suitable for MS systems
  • Includes fittings and restrictor
  • Replacement as required