Learn More
Julabo USA Inc 1m reinforced rubber tubing 1" ID
SDP
Catalog No. 507534146
Encompass_Preferred
Bath Accessories, Julabo, 1 m Reinforced rubber tubing, 1″ inner dia. (-40 … +120°C). For use with FL11006, FL20006, FL2506, FL4006, FL7006, FLW11006, FLW20006, FLW2506, FLW4006, FLW7006. Priced per meter.
Catalog No. 50-753-4146 Supplier Julabo USA Inc Supplier No. 8930325
Bath Accessories, Julabo, 1 m Reinforced rubber tubing, 1″ inner dia. (-40 … +120°C). For use with FL11006, FL20006, FL2506, FL4006, FL7006, FLW11006, FLW20006, FLW2506, FLW4006, FLW7006. Priced per meter.