Learn More
Julabo USA Inc 1m reinforced rubber tubing 3/4" ID
SDP
Catalog No. 507534145
Encompass_Preferred
Bath Accessories, Julabo, 1 m Reinforced rubber tubing, 3/4″ inner dia. (-40 … +120°C). For use with FL1203, FL1703, FL2503, FL4003, FLW1703, FLW2503, FLW4003. Priced per meter.
Catalog No. 50-753-4145 Supplier Julabo USA Inc Supplier No. 8930319
Bath Accessories, Julabo, 1 m Reinforced rubber tubing, 3/4″ inner dia. (-40 … +120°C). For use with FL1203, FL1703, FL2503, FL4003, FLW1703, FLW2503, FLW4003. Priced per meter.