Learn More
Electron Microscopy Sciences Rubis Reinforced Plastic Tweezers Style K35A, 120 mm OAL
SDP
Catalog No. 5023803
Encompass_Preferred
Made of PBTP (Polybutylenterephthalate) with 20% fiber-glass, the tweezers are resistant to chemicals and acids, highly isolating, and resists temperatures up to 200 degree C.
Tips: 120 mm
OAL: Flat, strong tips
Catalog No. 50-238-03 Supplier Electron Microscopy Sciences Supplier No. 7819301
Made of PBTP (Polybutylenterephthalate) with 20% fiber-glass, the tweezers are resistant to chemicals and acids, highly isolating, and resists temperatures up to 200 degree C.
Tips: 120 mm
OAL: Flat, strong tips