Learn More
MOLTOX Molecular Toxicology Inc Reinforced Clostridial Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948637
Encompass_Preferred
Reinforced Clostridial Agar is used for cultivating and enumerating clostridia and other anaerobes from food and clinical specimens. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-637 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S145
Reinforced Clostridial Agar is used for cultivating and enumerating clostridia and other anaerobes from food and clinical specimens. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.